Learn More
Iron(II) chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1023°C |
Melting Point | 674°C |
pH | 2.5 |
Physical Form | Powder |
Quantity | 5 g |
Linear Formula | FeCl2 |
UN Number | UN3260 |
Merck Index | 14,4043 |
Solubility Information | Freely soluble in water,alcohol,acetone;Soluble in water,alcohol and acetone; Slightly soluble in benzene; Insoluble in ether. |
Formula Weight | 126.75 |
Show More |
Safety and Handling
- Iron(II) chloride, anhydrous
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only