missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11312498
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
iron(2+);dibromide
[Fe++].[Br-].[Br-]
This item is not returnable. View return policy
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
iron(2+);dibromide
[Fe++].[Br-].[Br-]

Used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

Iron(II) bromide is used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 684°C
Physical Form Beads
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in water,THF,methanol,ethanol.
Formula Weight 215.67
Percent Purity 99.995%
Packaging Ampouled under argon
Sensitivity Air sensitive; Hygroscopic
Specific Gravity 4.636
Density 4.636
Chemical Name or Material Iron(II) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) bromide anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'