Learn More
Iron(II) bromide, 98%, anhydrous, Thermo Scientific Chemicals
Catalog Number p-3523467
491.85 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
[Fe++].[Br-].[Br-]
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
[Fe++].[Br-].[Br-]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 937°C
Melting Point 684°C
Color Black to Brown-Red
Physical Form Crystals or Powder
Quantity 10 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula FeBr2
Merck Index 15, 4069
Solubility Information Solubility in water: 1090g/L
IUPAC Name λ²-iron(2+) dibromide
Formula Weight 215.65
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Iron(II) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.