Få mere at vide
Novus Biologicals™ IRIS9 Plus Prestained Protein Ladder
Artikelnummer. 30165163
Skift visning
Mængde:
500 μl
Artikelnummer. Mængde
30165163 500 μl
1 options
Artikelnummer. 30165163 Leverandør Novus Biologicals™ Leverandørnr. NBP333167
 Begrænset vare
Forventet forsendelse: 25-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
Prestained Protein Ladder
The IRIS9 Plus Prestained Protein Ladder is a combination of 9 pre-stained proteins with molecular weights from 15 to 170 kDa. The 9 recombinant proteins are covalently coupled with blue chromophore, while 1 green band at 15 kDa, a red band at 70 kDa and a newly designed peacock green band at 60 kDa serve as reference bands. The IRIS9 Plus Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating the target protein size and validating the Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
TRUSTED_SUSTAINABILITY
Tekniske data
Formulering 20 mM Trisphosphate at pH 7.5, 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15% (v/v) Glycerol.
Til brug med (applikation) 3ul or 5ul per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5ul per well for general Western transferring.
Mængde 500 μl