Få mere at vide
Irinotecan Carboxylate Sodium Salt, Mikromol
Artikelnummer. 30460803
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30460803 25 mg
1 options
Artikelnummer. 30460803 Leverandør Mikromol Leverandørnr. MM1157.170025
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Irinotecan Carboxylate Sodium Salt, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Irinotecan Carboxylate Sodium Salt
CAS 1329502-92-2
Analyt- eller komponentnavne Irinotecan Carboxylate Sodium Salt
Forsendelsestilstand Room Temperature
Molekylær formel C33H39N4NaO7
Certificeringer/compliance ISO 9001
Synonym (alphaS)-2-[([1,4'-Bipiperidin]-1'-ylcarbonyl)oxy]-alpha,12-diethyl-9,11-dihydro-alpha-hydroxy-8-(hydroxymethyl)-9-oxo-indolizino[1,2-b]quinoline-7-acetic Acid Sodium Salt, CPT 11 Carboxylic Acid Sodium Salt;
Løsningstype Neat
SMIL [Na+].CCc1c2CN3C(=O)C(=C(C=C3c2nc4ccc(OC(=O)N5CCC(CC5)N6CCCCC6)cc14)[C@@](O)(CC)C(=O)[O-])CO
Molekylvægt (g/mol) 626.68