missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MedChemExpress iRGD peptide
Specifications
Specifications
| Renhed | 98.79% |
| CAS | 1392278-76-0 |
| Indhold og opbevaring | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Til brug med (applikation) | Cancer-programmed cell death |
| Molekylvægt (g/mol) | 948.04 g/mol |
| Produkttype | Peptides |
| Mængde | 10 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | H2O : ≥ 50 mg/mL (52.74 mM) |
| Synonym | c(CRGDKGPDC) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?