Få mere at vide
MedChemExpress iRGD peptide
Artikelnummer. 16422649
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16422649 1 mg
1 options
Artikelnummer. 16422649 Leverandør MedChemExpress Leverandørnr. HYP0122
MedChemExpress iRGD peptide is a 9-amino acid cyclic peptide, triggers tissue penetration of drugs by first binding to av integrins, then proteolytically cleaved in the tumor to produce CRGDK/R to interact with neuropilin-1, and has tumor-targeting and tumor-penetrating properties.
Tekniske data
Renhed 98.79%
CAS 1392278-76-0
Indhold og opbevaring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Til brug med (applikation) Cancer-programmed cell death
Molekylvægt (g/mol) 948.04 g/mol
Produkttype Peptides
Mængde 1 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : ≥ 50 mg/mL (52.74 mM)
Synonym c(CRGDKGPDC)