Få mere at vide
Irgarol-d9 (tert-butyl-d9), CDN
Artikelnummer. 30317057
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317057 0.01 g
30312722 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317057 Leverandør CDN Leverandørnr. CDND71470.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Irgarol-d9
CAS 1189926-01-9
Molekylær formel C11 D9 H10 N5 S
Mængde 0.01 g
Synonym N'-Cyclopropyl-N-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-methylsulfanyl-1,3,5-triazine-2,4-diamine, N2-Cyclopropyl-N4-[1,1-di(methyl-d3)ethyl-2,2,2-d3]-6-(methylthio)-1,3,5-triazine-2,4-diamine, Irgarol-d9, Irgarol-D9 (tert-butyl-D9)
InChI formel InChI=1 S/C11H19N5S/c1-11(2,3)16-9-13-8(12-7-5-6-7)14-10(15-9)17-4/h7H,5-6H2,1-4H3,(H2,12,13,14,15,16)/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])C(Nc1nc(NC2CC2)nc(SC)n1)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2-N-cyclopropyl-4-N-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 262.4225
Formel vægt 262.1926 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity