missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF7 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06987R
Additional Details : Weight : 0.00970kg
Description
IRF7 Polyclonal antibody specifically detects IRF7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDownSpecifications
IRF7 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDown | |
DyLight 550 | |
interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H | |
Antibody was raised against a peptide corresponding to 17 amino acids near the center of human IRF7. The immunogen is located within amino acids 200-230 of IRF7. | |
0.1 mL | |
Transcription Factors and Regulators | |
3665 | |
Store at 4°C in the dark. | |
IgG |