Learn More
Abnova™ IRF4/CEN6p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0183.200uL
Additional Details : Weight : 0.02000kg
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Store at 4°C in the dark. | |
FISH-Ce | |
Liquid |
3662 | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
200 μL |