Få mere at vide
Tocris Bioscience™ IRDye™ 680RD, NHS ester
Beskrivelse
Key information: IRDye™ 680RD, NHS ester is a near-infrared (NIR) fluorescent dye, supplied with an NHS ester reactive group for the labeling of primary amines.
Application: Fluorescence microscopy.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 680 nm and 694 nm, respectively.
Please see the protocol for further information on protein/antibody labeling and conjugation.
It is recommended to dissolve dye in water or DMSO (10 - 20 mg/mL)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sodium (Z)-2-((2Z,4E)-5-(5-Chloro-3,3-dimethyl-7-(3-sulfonatopropyl)-3H-pyrrolo[2,3-b]pyridin-7-ium-2-yl)-3-(3-(3-((2,5-dioxopyrrolidin-1-yl)oxy)-3-oxopropyl)phenyl)penta-2,4-dien-1-ylidene)-3,3-dimethyl-1-(3-sulfonatopropyl)indoline-5-sulfonate |
| CAS | 1373919-87-9 |
| Mængde | 500 μg |
| Mål | Fluorescent Dyes |
| Molekylær formel | C43H45ClN4Na2O13S3 |
| Renhed | 0.7 |