Få mere at vide
Irbesartan-d6 (benzyl-d6), CDN
Artikelnummer. 30314157
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314157 0.01 g
30311799 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314157 Leverandør CDN Leverandørnr. CDND75100.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Irbesartan-d6
CAS 2375621-21-7
Molekylær formel C25 D6 H22 N6 O
Mængde 0.01 g
Synonym Irbesartan D6 (Benzyl D6), Irbesartan-d6, 2-Butyl-3-[[2'-(1 H-tetrazol-5-yl)biphenyl-4-yl-2,3,5,6-d4]methyl-d2]-1,3-diazaspiro[4.4 ]non-1-en-4-one, 2-Butyl-3-[[2'-(2 H-tetrazol-5-yl)[1,1'-biphenyl-2,3,5,6-d4]-4-yl]methyl-d2]-1,3-diazaspiro[4.4 ]non-1-en-4-one
InChI formel InChI=1 S/C25H28N6O/c1-2-3-10-22-26-25(15-6-7-16-25)24(32)31(22)17-18-11-13-19(14-12-18)20-8-4-5-9-21(20)23-27-29-30-28-23/h4-5,8-9,11-14 H,2-3,6-7,10,15-17H2,1H3,(H,27,28,29,30)/i11D,12 D,13 D,14 D,17D2
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1c2ccccc2c3nnn[nH]3)C([2 H])([2 H])N4C(=NC5(CCCC5)C4=O)CCCC
IUPAC navn 2-butyl-3-[dideuterio-[2,3,5,6-tetradeuterio-4-[2-(1 H-tetrazol-5-yl)phenyl]phenyl]methyl]-1,3-diazaspiro[4.4 ]non-1-en-4-one
Molekylvægt (g/mol) 434.5664
Formel vægt 434.2701 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity