Få mere at vide
Tocris Bioscience™ IRAK1/4 Inhibitor I
Artikelnummer. 18768148
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18768148 10 mg
18740489 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18768148 Leverandør Tocris Bioscience™ Leverandørnr. 5665/10
IRAK4 and IRAK1 inhibitor
IRAK1/4 Inhibitor I is an IRAK4 and IRAK1 inhibitor (IC50 values are 0.2 and 0.3 μM, respectively).
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[1-[2-(4-Morpholinyl)ethyl]-1H-benzimidazol-2-yl]-3-nitrobenzamide
CAS 509093-47-4
Mængde 10 mg
Mål Other Kinase Inhibitors
Molekylær formel C20H21N5O4
Formel vægt 395.41
Renhed >98%