missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IQGAP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21443MFV610
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IQGAP1 Polyclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| IQGAP1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195RasGAP-like with IQ motifs, SAR1 | |
| Produced in rabbits immunized with E. coli-derived Human IQGAP1 fragment | |
| 0.1 mL | |
| Signal Transduction | |
| 8826 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion