Få mere at vide
Novus Biologicals IQGAP1 Antibody (2C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00008826-M01
Produktkode 18310699
  • 4405.00 DKK / 0.01mg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
IQGAP1 Monoclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, KnockDown
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
IQGAP1
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195RasGAP-like with IQ motifs, SAR1
IQGAP1 (NP_003861, 611 a.a. ~ 710 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. WQSNKDTQEAQKFALGIFAINEAVESGDVGKTLSALRSPDVGLYGVIPECGETYHSDLAEAKKKKLAVGDNNSKWVKHWVKGGYYYYHNLETQEGGWDEP
0.1 mg
Signal Transduction
8826
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, KnockDown
2C5
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Knockdown Validated
NP_003861
Mouse
IgG purified
RUO
Primary
Human
Purified