Learn More
Memmert™ IQ/OQ Document with Test Data for IN450
Product Code. 17547725 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IN450 Incubator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17547725 IN450 Incubator Each
1 options
Product Code. 17547725 Supplier Memmert™ Supplier No. IN450D00127
 Restricted Item
Forventet forsendelse: 21-12-2026
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
For Use With (Equipment) IN450 Incubator
Product Type IQ/OQ Documentation
Includes Test Data