Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
For one free-selectable temperature and humidity value, and measuring of light intensity
Brand:  Memmert™ HPP110ecoD00137
 View more versions of this product
Product Code. 17129684
  • 31940.00 DKK / Each
This item is not returnable. View return policy
Description
Description
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
Specifications
HPP110eco
IQ/OQ Document with Device-specific Works Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity > HPP110eco