Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature and Humidity
Product Code. 17199714 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17199714 HPP1400eco Each
17109644 HPP1060eco Each
17109824 HPP410eco Each
17119684 HPP110eco Each
17139744 HPP2200eco Each
17159774 HPP260eco Each
17189864 HPP750eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17199714 Supplier Memmert™ Supplier No. HPP1400ecoD00136
 Restricted Item
Forventet forsendelse: 21-12-2026
For one free-selectable temperature and humidity value, Includes temperature distribution survey at Memmert to DIN 12880:2007-05
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
For Use With (Equipment) HPP1400eco
Product Type IQ/OQ Document with Device-specific Works Test Data