Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
Brand:  Memmert™ HPP2200ecoD00127
 View more versions of this product
Product Code. 17129744
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
Specifications
HPP2200eco
IQ/OQ Document with Device-specific Works Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature > HPP2200eco