Få mere at vide
Iprodione-d5, TRC
Artikelnummer. 30407824
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30407824 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407824 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7450022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iprodione-d5
CAS 1215631-57-4
Molekylær formel C13 2H5 H8 Cl2 N3 O3
Mængde 2.5 mg
Synonym Iprodione D5 (dichlorophenyl D3 hydantoin D2)
InChI formel InChI=1S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7H,6H2,1-2H3,(H,16,20)/i3D,4D,5D,6D2
SMIL [2H]c1c(Cl)c([2H])c(N2C(=O)N(C(=O)NC(C)C)C([2H])([2H])C2=O)c([2H])c1Cl
IUPAC navn 5,5-dideuterio-3-(3,5-dichloro-2,4,6-trideuteriophenyl)-2,4-dioxo-N-propan-2-ylimidazolidine-1-carboxamide
Molekylvægt (g/mol) 335.2
Formel vægt 334.0648