Få mere at vide
Iprodione-d5 (3,5-dichlorophenyl-2,4,6-d3; hydantoin-5,5-d2), CDN
Artikelnummer. 30319784
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319784 0.005 g
30310366 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319784 Leverandør CDN Leverandørnr. CDND64220.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iprodione-d5
CAS 1215631-57-4
Molekylær formel C13 2H5 H8 Cl2 N3 O3
Mængde 0.005 g
InChI formel InChI=1 S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7 H,6H2,1-2H3,(H,16,20)/i3D,4 D,5 D,6D2
SMIL [2 H]c1c(Cl)c([2 H])c(N2C(=O)N(C(=O)NC(C)C)C([2 H])([2 H])C2=O)c([2 H])c1Cl
IUPAC navn 5,5-dideuterio-3-(3,5-dichloro-2,4,6-trideuteriophenyl)-2,4-dioxo-N-propan-2-ylimidazolidine-1-carboxamide
Molekylvægt (g/mol) 335.2
Formel vægt 334.0648 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity