Få mere at vide
Ipratropium Bromide, TRC
Artikelnummer. 30133464
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133464 2.5 g 2.5g
17994375 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133464 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7404952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22254-24-6
Molekylær formel C20H30NO3 . Br
Molekylvægt (g/mol) 412.36
Synonym 3α-Hydroxy-8-isopropyl-1αH,5αH-tropanium Bromide (±)-Tropate, (3-Endo,8-syn)-3-(3-Hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane Bromide, (endo,syn)-(±)-3-(3-Hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane Bromide, 8-Isopropylatropinium Bromide, (endo,syn)-3-(3-Hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane Bromide, Atrovent, Atrovent Forte, N-Isopropylatropinium Bromide, N-Isopropylnoratropinium Bromomethylate, Sch 100, Sch 1000
IUPAC navn 3-((3-hydroxy-2-phenylpropanoyl)oxy)-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium bromide
SMIL OCC(C1=CC=CC=C1)C(O[C@H]2C[C@H]3CC[C@@H](C2)[N@@+]3(C)C(C)C)=O.[Br-]
Tekniske data
Kemisk navn eller materiale Ipratropium Bromide
Mængde 2.5 g
InChI formel InChI=1S/C20H30NO3.BrH/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H/q+1;/p-1
Formel vægt 411.14