Få mere at vide
Ipratropium Bromide Monohydrate, TRC
Artikelnummer. 30134854
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134854 500 mg
30009864 50 mg
17981035 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134854 Leverandør Toronto Research Chemicals Leverandørnr. TRCI740500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66985-17-9
Molekylær formel C20H30NO3.Br.H2O
Molekylvægt (g/mol) 430.38
Synonym 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide, hydrate (1:1:1), (3-endo,8-syn)-, 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide, monohydrate, (3-endo,8-syn)- (9CI), 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide, monohydrate, (endo,syn)-(±)-, 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide, monohydrate, (endo,syn)-, Ipratropium bromide monohydrate
IUPAC navn (1R,3r,5S,8r)-3-((3-hydroxy-2-phenylpropanoyl)oxy)-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium bromide hydrate
SMIL OCC(C1=CC=CC=C1)C(O[C@H]2C[C@H]3CC[C@@H](C2)[N@@+]3(C)C(C)C)=O.[Br-].O
Tekniske data
Kemisk navn eller materiale Ipratropium Bromide Monohydrate
Mængde 500 mg
InChI formel InChI=1S/C20H30NO3.BrH.H2O/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H;1H2/q+1;;/p-1/t16-,17+,18+,19?,21+;;
Formel vægt 429.15