Få mere at vide
Ipratropium Bromide Monohydrate, LoGiCal
Artikelnummer. 30457659
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457659 10 mg
1 options
Artikelnummer. 30457659 Leverandør LoGiCal Leverandørnr. LGCFOR1249.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ipratropium Bromide Monohydrate
CAS 66985-17-9
Analyt- eller komponentnavne Ipratropium Bromide Monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C20H32BrNO4
Certificeringer/compliance ISO 9001
Synonym (3-endo,8-syn)-3-(3-Hydroxy-1-oxo-2-phenylpropyl)-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane Bromode Monohydrate, Atropine Isopropyl Bromide;
InChI formel InChI=1S/C20H30NO3.BrH.H2O/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H;1H2/q+1;;/p-1/t16-,17+,18+,19?,21+;;
Løsningstype Neat
SMIL OCC(C1=CC=CC=C1)C(O[C@H]2C[C@H]3CC[C@@H](C2)[N@@+]3(C)C(C)C)=O.[Br-].O
IUPAC navn (1R,3r,5S,8r)-3-((3-hydroxy-2-phenylpropanoyl)oxy)-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium bromide hydrate
Industry Type Clinical Testing
Molekylvægt (g/mol) 430.38
Mængde 10 mg
Formel vægt 429.15 g/mol