Få mere at vide
MedChemExpress Ipilimumab
Artikelnummer. 16403069
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16403069 1 mg
1 options
Artikelnummer. 16403069 Leverandør MedChemExpress Leverandørnr. HYP9901
MedChemExpress Ipilimumab is a fully human monoclonal IgG1κ antibody against the cytotoxic T-lymphocyte antigen-4 (CTLA-4), an immune-inhibitory molecule expressed in activated T cells and in suppressor T regulatory cells.
Tekniske data
Antigen Others
Mængde 1 mg
Indhold og opbevaring Please store the product under the recommended conditions in the Certificate of Analysis.
Produkttype Inhibitory Antibodies
Forsendelsestilstand Shipping with dry ice
CAS 477202-00-9
Renhed eller kvalitetsklasse 99.0%