Få mere at vide
Tocris Bioscience™ IP7e
Potent Nurr1 activator
Brand: Tocris Bioscience™ 5699/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IP7e is a potent Nurr1 activator (EC50 = 3.9 nM). Delays onset and reduces severity of symptoms in mice with experimental autoimmune encephalomyelitis (EAE). Also suppresses NF-κB signaling. Brain penetrant and orally bioavailable.Tekniske data
| 6-[4-[(2-Methoxyethoxy)methyl]phenyl]-5-methyl-3-phenyl-isoxazolo[4,5-c]pyridin-4(5H)-one | |
| 500164-74-9 | |
| 50 mg |