missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ IOX 2

Artikelnummer. 18024822
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18024822 10 mg
18064972 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18024822 Leverandør Tocris Bioscience™ Leverandørnr. 4451/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor

Potent inhibitor of HIF-1α prolyl hydroxylase-2 (PHD2) (IC50 = 21 nM). Displays over 100-fold selectivity for PHD2 over factor inhibiting HIF-1 (FIH-1) and histone demethylases JMJD2A, JMJD2C, JMJD2E and JMJD3. Cell permeable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931398-72-0
Molekylær formel C19H16N2O5

Specifications

Kemisk navn eller materiale IOX 2
Mængde 10 mg
Formel vægt 352.34
Renhed >99%
Molekylvægt (g/mol) 352.346
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.