Få mere at vide
Iopydone, LoGiCal
Artikelnummer. 30463721
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30463721 5 mg
1 options
Artikelnummer. 30463721 Leverandør LoGiCal Leverandørnr. LGCFOR3116.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iopydone
CAS 5579-93-1
Analyt- eller komponentnavne Iopydone
Forsendelsestilstand Room Temperature
Molekylær formel C5 H3 I2 N O
Certificeringer/compliance ISO 9001
Synonym 4(1H)-Pyridinone, 3,5-diiodo-, 3,5-Diiodo-4(1H)-pyridone, 3,5-Diiodo-4-pyridone, DJP, Iopydon, Iopydone, NSC 135284
InChI formel InChI=1S/C5H3I2NO/c6-3-1-8-2-4(7)5(3)9/h1-2H,(H,8,9)
Løsningstype Neat
SMIL IC1=CNC=C(I)C1=O
IUPAC navn 3,5-diiodo-1H-pyridin-4-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 346.89
Mængde 5 mg
Formel vægt 346.8304 g/mol