Få mere at vide
Iopromide-d3, TRC
Artikelnummer. 30406847
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30406847 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406847 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7357020.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iopromide-d3
CAS 1189947-73-6
Renhedsgrad noter HPLC
Molekylær formel C18H21D3I3N3O8
Mængde 0.5 mg
Synonym Iopromide D3 (methoxy D3)
InChI formel InChI=1S/C18H24I3N3O8/c1-24(4-9(28)6-26)18(31)12-13(19)11(17(30)22-3-8(27)5-25)14(20)16(15(12)21)23-10(29)7-32-2/h8-9,25-28H,3-7H2,1-2H3,(H,22,30)(H,23,29)/i2D3
SMIL [2H]C([2H])([2H])OCC(=O)Nc1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)N(C)CC(O)CO)c1I
Molekylvægt (g/mol) 794.13
Procent renhed >95