Få mere at vide
Ion Chromatography Standard: Dichloroacetate @ 500 μg/mL in H2O, VHG™
Artikelnummer. 17927824
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17927824 100 ml
17960065 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17927824 Leverandør VHG Labs Leverandørnr. VHGIDCA500100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Ion Chromatography Standard: Dichloroacetate @ 500 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dichloroacetate
CAS 13425-80-4
Metoder Ion Chromatography
Analyt- eller komponentnavne Dichloroacetate
Forsendelsestilstand Room Temperature
Molekylær formel C2 H Cl2 O2
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17025
Synonym Acetic acid, 2,2-dichloro-, ion(1-), Acetic acid, dichloro-, ion(1-) (8CI,9CI), 2,2-Dichloroacetate, Dichloroacetate anion, Dichloroacetate ion, Dichloroacetate ion(1-)
InChI formel InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/p-1
SMIL [O-]C(=O)C(Cl)Cl
IUPAC navn 2,2-dichloroacetate
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 127.935
Mængde 100 ml
Formel vægt 126.9359 g/mol