Learn More
Instrument Check Standard 6 For Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ICMIX6-100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
HPO4-2 | 150ÎĽg/mL |
SO4-2 | 150ÎĽg/mL |
Br- | 100ÎĽg/mL |
Cl- | 50ÎĽg/mL |
NO2-N | 25ÎĽg/mL |
NO3-N | 25ÎĽg/mL |
F- | 20ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Instrument Check Standard 6 | |
With Certificate of Analysis | |
Ion Chromatography | |
Standard Solution | |
H2O | |
Components in water: Br- •100μg/mL Cl- •50μg/mL F- •20μg/mL HPO4-2 •150μg/mL NO3--N •25μg/mL NO2--N •25μg/mL SO4-2 •150μg/mL |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Certified Reference Material | |
125 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product intended for laboratory use only