missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorate Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
775.00 DKK - 1515.00 DKK
Specifications
Certificate of Analysis | With Certificate of Analysis |
---|---|
CAS Min % | 0.1, 99.9 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
CAS Max % | 0.1, 99.9 |
CAS | 7732-18-5 |
Technique | Ion Chromatography |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Solution Type | Standard Solution |
Type | Certified Reference Material |
Solvent or Matrix | H2O |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10162002
|
SPEX Certiprep
AS-CLO39-2Y |
125 mL | Triple-leached LDPE bottle | N/A | ||||||
11486098
|
SPEX Certiprep
AS-CLO39-2X |
500 mL | Triple-leached LDPE bottle |
1515.00 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
ClO3- | 1000ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
With Certificate of Analysis | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
7732-18-5 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Certified Reference Material | |
Chlorate Standard |
0.1, 99.9 | |
0.1, 99.9 | |
Ion Chromatography | |
Standard Solution | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chlorate Standard for Ion Chromatography, SPEX CertiPrep™