Få mere at vide
Iomeprol-d3, TRC
Artikelnummer. 30405768
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30405768 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405768 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7305021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iomeprol-d3
CAS 1185146-41-1
Molekylær formel C17H19D3I3N3O8
Mængde 1 mg
Synonym Iomeprol D3, Iomeprol D3 (N-methyl D3)
InChI formel InChI=1S/C17H22I3N3O8/c1-23(9(29)6-26)15-13(19)10(16(30)21-2-7(27)4-24)12(18)11(14(15)20)17(31)22-3-8(28)5-25/h7-8,24-28H,2-6H2,1H3,(H,21,30)(H,22,31)
SMIL [2H]C([2H])([2H])N(C(=O)CO)c1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I
Molekylvægt (g/mol) 780.1