Learn More
Iodoethanol, TRC
Product Code. 30137187
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137187 1 g 1g
30134282 250 mg 250mg
30010650 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30137187 Supplier Toronto Research Chemicals Supplier No. TRCI7058751G
 Restricted Item
Forventet forsendelse: 03-12-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 624-76-0
Molecular Formula C2 H5 I O
Molecular Weight (g/mol) 171.965
Synonym 2-Iodoethanol, 2-Hydroxy Iodide, 2-Iodoethan-1-ol, Ethylene Iodohydrin, NSC 85227
IUPAC Name 2-iodoethanol
SMILES OCCI
Specifications
Quantity 1 g
InChI Formula InChI=1S/C2H5IO/c3-1-2-4/h4H,1-2H2
Formula Weight 171.939
Chemical Name or Material Iodoethanol