Få mere at vide
Iodoethane-2-d1, CDN
Artikelnummer. 30312700
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312700 0.25 g
30319787 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312700 Leverandør CDN Leverandørnr. CDND14820.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Iodoethane-2-D
CAS 57460-45-4
Molekylær formel C2 D H4 I
Mængde 0.25 g
Synonym Ethane-d, 2-iodo- (9 CI), [2-2H1]Ethyl iodide, 2-Iodoethane-d, 1-Iodoethane-2-d, Ethyl iodide-2-d, Ethyl-2-d1 iodide, 1-Iodoethane-2-D
InChI formel InChI=1 S/C2H5I/c1-2-3/h2H2,1H3/i1D
SMIL [2 H]CCI
IUPAC navn 1-deuterio-2-iodoethane
Molekylvægt (g/mol) 156.9717
Formel vægt 156.9499 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity