Få mere at vide
Iodoethane-2,2,2-d3, CDN
Artikelnummer. 30319315
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319315 1 g
30310059 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319315 Leverandør CDN Leverandørnr. CDND05371G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1,1-trideuterio-2-iodoethane
CAS 7439-87-4
Molekylær formel C2 2H3 H2 I
Mængde 1 g
InChI formel InChI=1 S/C2H5I/c1-2-3/h2H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CI
IUPAC navn 1,1,1-trideuterio-2-iodoethane
Molekylvægt (g/mol) 158.984
Formel vægt 158.962 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity