Learn More
Iodine Trichloride, Sigma-Aldrich™
Product Code. 15854161
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854161 100 g 100g
15844161 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15854161 Supplier Sigma Aldrich Supplier No. 8047720100
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 865-44-1
Specifications
Melting Point >60°C (decomposition)
Quantity 100 g
Density 3.12 g/cm3 at 20°C
MDL Number MFCD00036291
Synonym Iodine trichloride, Iodine trichloride
Molecular Weight (g/mol) 233.26
Grade Research
Chemical Name or Material Iodine Trichloride