Learn More
Iodine Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Brand: Fisher Chemical J/4400/PB08
Additional Details : CAS Number : 7553-56-2 Weight : 0.50000kg
Quantity | 500mL |
---|
Chemical Identifiers
7553-56-2 | |
253.809 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
molecular iodine |
I2 | |
11355 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 | |
II |
Specifications
7553-56-2 | |
11355 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
Liquid | |
100°C | |
500mL |
I2 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.809 | |
CHEBI:17606 | |
Plastic Coated Glass bottle | |
Brown |