Få mere at vide
Iodide (from NaI) Standard: I- @ 10000 μg/mL in H2O, VHG™
Artikelnummer. 30002973
Skift visning
Mængde:
500 ml
Artikelnummer. Mængde
30002973 500 ml
1 options
Artikelnummer. 30002973 Leverandør VHG Labs Leverandørnr. VHGI1PI500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Iodide (from NaI) Standard: I- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iodide
CAS 20461-54-5
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Iodide
Forsendelsestilstand Room Temperature
Molekylær formel I
Synonym I−
InChI formel InChI=1S/HI/h1H/p-1
SMIL [I-]
IUPAC navn iodide
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 126.904
Mængde 500 ml
Formel vægt 126.904 g/mol