Få mere at vide
Invitrogen™ Zero Background™ Cloning Kit
Mængde:
25 μg
Pakningsstørrelse:
25µg
Beskrivelse
Indeholder
25μg supercoiled pZErO-1 (the Zero Background Cloning Kit)or pZErO-2 vector (the Zero Background/Kan Cloning Kit) in TE, 200 ng blunt-end test inserts, TOP10 E. coli stab, 25μL T4 DNA ligase, 10X ligation buffer, 1mL sterile water, and 2mL sterile TE. The Zero Background Cloning Kit also contains 1.25mL Zeocin selection agent.
- pZErO vectors use positive selection to eliminate high background
- pZErO-1 vector takes advantage of small size of the Zeocin resistance gene to reduce overall vector size for increased ligation and transformation efficiencies
- pZErO-2 vector includes the kanamycin resistance gene
Cloning, PCR Cloning
Tekniske data
Tekniske data
| Bakterie- eller gærstamme | TOP10 |
| Indhold og opbevaring | Store TOP10 stab at room temperature. Store the remaining kit components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Mængde | 25 μg |
| Vektor | TA Cloning Vectors, pCR |