Få mere at vide
Invitrogen™ T-REx™ Complete Kit, with pcDNA™4/TO© Vector
Artikelnummer. 10624033
Skift visning
Mængde:
1 kit
Pakningsstørrelse:
5g
Artikelnummer. Mængde unitSize
10624033 1 kit 5g
1 options
Regulatory: A license is required to purchase this product; ROYAL ORDER OF APRIL 12, 1974.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 10624033 Leverandør Invitrogen™ Leverandørnr. K102001
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: A license is required to purchase this product; ROYAL ORDER OF APRIL 12, 1974.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Indeholder
20μg of pcDNA4/TO or 20μg each of pcDNA4/TO/myc-His A, B, and C; a positive control vector, pcDNA;6/TR, and forward and reverse sequencing primers plus 1g Zeocin (100mg/mL), 50mg of Blasticidin, and 5g of tetracycline. All vectors are supercoiled and provided lyophilized.
Regulated mammalian expression system yields exceptionally high levels of induced expression
  • Uses complete CMV promoter and adds control elements from bacterial tetracycline resistance operon to effectively repress and derepress transcription from one of strongest mammalian promoter sequences known
  • Specific Activation: uses a repressor mechanism that blocks transcription from powerful CMV promoter in absence of tetracycline
  • Because elements do not use viral transactivators, you can achieve high-level expression from complete CMV promoter without secondary, non-specific activation of host genes
  • T-REx transcriptional control elements are provided through two tetracycline operator sequences (TetO2) that have been inserted between TATA box of CMV promoter and transcriptional start site
  • TetO2 sequence itself has no effect on expression
  • When the tetracycline repressor protein (TR) is present, it effectively binds TetO2 sites and blocks transcription initiation
  • Tetracycline added to culture medium binds to, and changes conformation of, TR protein; This change causes TR protein to release TetO2 sites, derepressing transcription from CMV promoter
  • Result is high-level expression of gene of interest
  • Expression levels can be modulated based on tetracycline concentration and can be induced to levels that are achieved with constitutive CMV expression vectors
  • T-REx is a powerful inducible mammalian expression system that allows to regulate expression from complete human cytomegalovirus (CMV) enhancer-promoter
  • Complete CMV enhancer-promoter sequence containing two copies of tetracycline operator TetO2 sequence for high-level regulated expression
  • Zeocinor hygromycin resistance gene for effective selection of stable mammalian cell lines
  • Large multiple cloning site to simplify cloning
  • In addition, pcDNA™4/TO/myc-His offers a c-myc epitope for rapid detection of the recombinant protein with an Anti-myc Antibody and a polyhistidine (6xHis) sequence for simple purification of recombinant protein with nickel-chelating resin and detection with Anti- His(C-term) Antibody
  • Regulatory vector, pcDNA™6/TR, is provided for high-level expression of the tetracycline repressor (TR) protein
  • Expresses Blasticidin resistance gene for rapid selection of mammalian cell lines that stably express the TR protein
Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
TRUSTED_SUSTAINABILITY
Tekniske data
Konstitutivt eller inducerbart system Inducible
Leveringstype Transfection
Inducerende middel Tetracycline
Promotor CMV/TO
Produkttype T-Rex Complete Cloning Kit with Vector
Udvælgelsesagent (eukaryot) Zeocin™, Blasticidin
Indhold og opbevaring The T-REx™ Core Kit includes 20 μg of pcDNA™4/TO, a positive control vector, pcDNA™6/TR, and forward and reverse sequencing primers.
The T-REx™ Complete Kit includes the Core Kit plus 1 g Zeocin™ (100 mg/ml), 50 mg of Blasticidin, and 5 g of tetracycline.
All vectors are supercoiled and provided lyophilized.
Store all components at -20°C. Zeocin™ and tetracycline should be protected from exposure to light. All components are guaranteed stable for 6 months when properly stored.
Protein tag Untagged
Kloningsmetode Restriction Enzyme/MCS
Mængde 1 kit
Vektor pcDNA
Produktlinje T-REx, pcDNA
Til brug med (applikation) Regulated Expression
Vis mere Vis mindre
For Research Use Only. Not for use in diagnostic procedures.