Learn More
Invitrogen™ Platinum™ II Hot-Start PCR Master Mix (2X)
Invitrogen™ Hot-Start PCR Master Mix for fast and easy PCR
642.00 DKK - 11390.00 DKK
Specifications
Format Tube
GC-Rich PCR Performance High
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15889844
View Documents Promotion Details
Invitrogen™
14000012
50 Reactions
642.00 DKK
50 reactions
View alternative products
Save Up to 
»
In Stock
to see stock.
15899844
View Documents Promotion Details
Invitrogen™
14000013
200 Reactions
2480.00 DKK
200 reactions
View alternative products
Save Up to 
»
In Stock
to see stock.
15809854
View Documents Promotion Details
Invitrogen™
14000014
1000 Reactions
11390.00 DKK
1000 reactions
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
Invitrogen™ Platinum™ Taq II Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq, and leading hot-start technology.
  • Innovative buffer—enables universal primer annealing protocol by isostabilizing primer-template duplex structures
  • Engineered Taq—confers  fast cycling and  tolerance to common inhibitors
  • Platinum hot-start technology—ensures superior specificity, sensitivity, and yields; allows for room temperature reaction setup
  • Optional green buffer—reduces pipetting error with direct gel loading
Additional:
  • Antibody-mediated hot-start
  • Universal annealing protocool
  • Inhibitor resistance
Applications:
  • Genotyping
  • High-throughput PCR
  • GC-rich templates
  • Fast PCR
  • Amplification of samples with suboptimal purity
  • Direct PCR with blood
  • PCR from DNA extracted from FFPE samples
  • Colony PCR
  • PCR from bacterial 16S rRNA genes
Specifications
Specifications
Tube
High