Learn More
Invitrogen™ Gateway™ pDEST™8 Vector
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
Brand: Invitrogen™ 11804010
Additional Details : Weight : 0.02000kg
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Description
- Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Baculovirus Production and Titering, Cloning, Gateway Cloning, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Polyhedrin | |
All destination vectors are provided lyophilized and supercoiled. | |
Gateway | |
Gateway™ |
Expression Vector | |
Untagged | |
pDEST, Gateway Baculovirus Vectors |
For Research Use Only. Not for use in diagnostic procedures.