missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chargeswitch™ Pro Filter Plasmid Midi Kit

The ChargeSwitch™-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.

Brand:  Invitrogen™ CS31104

Additional Details : Weight : 2.03000kg

Product Code. 10756675

  • 3875.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products

Includes: The ChargeSwitch™-Pro Plasmid Kit contains Columns with Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer.

Description

Description

The ChargeSwitch™-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications. The system is based upon a novel dual/nested column design which facilitate the fastest and simplest protocol for high quality plasmid. The system is composed of two columns. The inner flat bottomed column, also referred to as “lysate clarification” column, contains filter materials suitable for rapid clarification of bacterial lysate. The outer luer bottomed column, also referred to as “binding” column, contains ChargeSwitch™ (CST) derivatized membrane that immediately binds plasmid DNA from the clarified lysate. This nested-column design enables simultaneous clarification and DNA binding in a single short centrifugation (or vacuum) step. The combined advantages of novel, optimized plastics and a unique chemistry are an extreme competitive advantage of speed and convenience.

  • 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
  • Cleaner purified product for improved performance in downstream applications
  • A novel Dual/nested column design for simultaneous lysate clearing and binding step which offers the fastest and the simplest protocol
  • Endotoxin units less than 10 EU/μg DNA for
  • Vacuum Elution can be done with the EveryPrep™ Universal Vacuum Manifold (Cat. No. K211101)

How it works
The ChargeSwitch™ technology replaces the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification with a simple ion-exchange mechanism. At low pH values (less than 6.5) the ChargeSwitch™ surface ligand is positively charged and binds plasmid DNA. Raising the pH (>8.5) neutralizes the charge and elutes the bound nucleic acid. The ChargeSwitch™ protocol retains the ease and familiarity of the common silica spin column protocol with a binding, washing, and elution step, making adapting to the ChargeSwitch™ method effortless.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Spin Column
Spin Column
Midi
0.5 to 1 mL
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
Columns with Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer
ChargeSwitch™
25 Preps
Room Temperature
ChargeSwitch™
200 μg
The ChargeSwitch™-Pro Plasmid Kit contains Columns with Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer. Store at room temperature. Guaranteed stable for 6 months when properly stored.
ChargeSwitch Chemistry
Bacterial Culture
Plasmid DNA
Not High-throughput Compatible (Manual)
100-200 μg (Medium-Scale) Plasmid DNA
Pro Filter Plasmid Midi Kit
Transfection Grade
Plasmid DNA
30 min.
25 ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.