Beskrivelse
- Comprised of disposable glass without ring mark.
- Complies with DIN ISO 7550.
- CE Marked in accordance with IVD-Directive 98/79
- Calibrated to contain (TC, In.)
- Volume is defined by end-to-end filling
- Volumes greater than 5μL include DE-M marking.
Tekniske data
Tekniske data
| Nøjagtighed | ≤±0.5% |
| Kalibrering | To Contain |
| Certificeringer/compliance | DIN ISO 7550, IVD-Directive 98/79 CE |
| Materiale | Glass |
| Mængde | 250/Box |
| Volumen (metrisk) | 1 μL |
| Længde (metrisk) | 32 mm |
| Engangs | Single-use |