Få mere at vide
Thermo Scientific Chemicals Interleukin 3, mouse
Artikelnummer. 15428639
Skift visning
Mængde:
10 μg
2 μg
Pakningsstørrelse:
10µg
2µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15428639 2 μg 2µg
15418639 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15428639 Leverandør Thermo Scientific Chemicals Leverandørnr. J67104.&E
 Begrænset vare
Forventet forsendelse: 07-12-2026
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Tekniske data
Molekylvægt (g/mol) 15.6 kd
Mængde 2 μg
Emballage Plastic bottle
Form Lyophilized