missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Integrin beta 8 Antibody (416922), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4775W
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry, CyTOF
Tekniske data
| Integrin beta 8 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 416922 | |
| Flow Cytometry, CyTOF-ready | |
| integrin beta-8, integrin, beta 8 | |
| Mouse myeloma cell line NS0-derived recombinant human Integrin beta 8, Glu43-Arg684 (predicted), Accession # P26012 | |
| 0.1 mL | |
| Signal Transduction | |
| 3696 | |
| Store at 4°C in the dark. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion