Beskrivelse
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Tekniske data
Tekniske data
| Antigen | Integrin alpha E/CD103 |
| Applikationer | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Klassifikation | Monoclonal |
| Klon | Ber-ACT8 |
| Konjugeret | FITC |
| Fortynding | Western Blot 1:100-1:2000, Flow Cytometry 4ul reagent 100 ul of whole blood or 10^6 cells in a suspension., Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Gene Alias | antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen |
| Gensymboler | ITGAE |
| Værtsarter | Mouse |
| Immunogen | HTLV-1 induced human T cell line MAPS16 |
| Vis mere |
For Research Use Only