missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Integrin alpha 5 beta 1 Antibody (BMS patent anti-Integrin alpha5beta1), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28423CL1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
Integrin alpha 5 beta 1 | |
Monoclonal | |
CoraFluor 1 | |
CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A | |
Integrin a5b1 (ITGA5 & ITGB1) | |
0.1 mL | |
Angiogenesis, Cancer, Immunology | |
3678 | |
Store at 4°C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry, ELISA, Functional Assay | |
BMS patent anti-Integrin alpha5beta1 | |
PBS | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Integrin alpha 5 beta 1 Antibody (BMS patent anti-Integrin alpha5beta1), CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1
Ser du en mulighed for forbedring?Del en indholdskorrektion