missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Integrin alpha 2b beta 3 Antibody (10E5), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81080AF647
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Tekniske data
| Integrin alpha 2b beta 3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3674 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 1000000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| ITGA2B | |
| Human blood platelets. | |
| 0.1 mL | |
| Primary | |
| The antibody binds to Integrin alpha 2b beta 3 with an affinity (Kd) of 16.2+-6 nM. The antibody completely inhibits platelet aggregation at 7.8 ug/ml. | |
| Store at 4°C in the dark. | |
| IgG κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion